Anti-Staufen Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # 76120-262
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Staufen
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Horse,Chicken,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Epitope:421-520/577
- Form:Liquid
- Gene ID:6780
- Antigen Synonyms:PPP1R150|STAU1|STAU|Double-stranded RNA-binding protein Staufen homolog 1
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:63 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Staufen
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76120-262
- Packaging:Vial
Specifications
About this item
Binds double-stranded RNA (regardless of the sequence) and tubulin. May play a role in specific positioning of mRNAs at given sites in the cell by cross-linking cytoskeletal and RNA components, and in stimulating their translation at the site.
Type: Primary
Antigen: Staufen
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 421-520/577
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken, Horse, Others