Specifications
- Antibody Type:Primary
- Antigen Name:RELM beta
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Epitope:24-80/111
- Form:Liquid
- Gene ID:84666
- Antigen Synonyms:XCP2|Colon carcinoma-related gene protein|RETNL2. RETNLB|CCGR|RELM beta|Colon carcinoma related gene protein|Cysteine rich secreted protein A12 alpha like 1|Resistin like beta|HXCP2|RETNB_HUMAN|Resistin like protein beta|Colon and small intestine-specic cysteine-rich protein|FIZZ1|Cysteine-rich secreted protein FIZZ2|RELMb|Cysteine rich secreted protein FIZZ2|Cysteine-rich secreted protein A12-alpha-like 1|FIZZ2|Found in Inflammatory Zone 2|Resistin Like Protein Beta|RELMbeta|Resistin-like beta|C/EBP epsilon regulated myeloid specic secreted cysteine rich protein precursor 2|Found in inflammatory zone 1|Cysteine rich secreted A12 alpha like protein 1|Colon and small intestine specic cysteine rich protein
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:9 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human RELM beta/CCGR
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76081-156
- Packaging:Vial
Specifications
About this item
The cysteine-rich, adipose tissue-specific, secretory factor resistin (resistance to insulin, also known as ADSF) is a secreted hormone that potentially links obesity to diabetes. Resistin is rich in serine and cysteine residues and contains a unique cysteine repeat motif. Resistin and the resistin-like molecules share the characteristic cysteine composition and other signature features. Resistin-like a is a secreted protein that has restricted tissue distribution and is most highly expressed in adipose tissue. Another family member, Resistin-like b, is a secreted protein expressed only in the gastrointestinal tract, particularly in the colon, in both mouse and human. Resistin-like b expression is highest in proliferative epithelial cells and is markedly increased in tumors, suggesting a role in intestinal proliferation.
Type: Primary
Antigen: RELM beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 24-80/111
Host: Rabbit
Isotype: IgG
Reactivity: Human