Anti-14-3-3(Thr232) Rabbit Polyclonal Antibody (Alexa Fluor® 680)
76080-790
: Bioss
- Antibody Type:Primary
- Antigen Name:14-3-3(Thr232)
- Antigen Symbol:14-3-3(Thr232)
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:7531
- Antigen Synonyms:Tyrosine 3 monooxygenase/tryptophan 5 monooxygenase activation protein, tyrosine 3/tryptophan 5 -monooxygenase activation protein|Protein kinase C inhibitor protein1|14-3-3, mitochondrial import stimulation factor L subunit|protein kinase C inhibitor protein-1
- Modification:Phosphorylation (Thr232)
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:29 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human 14-3-3 around the phosphorylation site of Thr232
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76080-790
- Packaging:Vial
Adapter protein implicated in the regulation of a large spectrum of both general and specialized signaling pathway. Binds to a large number of partners, usually by recognition of a phosphoserine or phosphothreonine motif. Binding generally results in the modulation of the activity of the binding partner.
Type: Primary
Antigen: 14-3-3(Thr232)
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human