Anti-Matrin 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # 76079-818
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Matrin 3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Size:100 µl
- Epitope:421-520/847
- Form:Liquid
- Gene ID:9782
- Antigen Synonyms:KIAA0723|MATR3_HUMAN|Matrin3|MATR3|VCPDM|Matrin-3|MPD2|Matr3
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:95 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Matrin 3
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76079-818
- Packaging:Vial
Specifications
About this item
Matrin 3 (MATR 3) is an internal nuclear matrix protein that may play a role in transcription or may interact with other nuclear matrix proteins to form the internal fibrogranular network. In association with the SFPQ-NONO heteromer MATR 3 may play a role in nuclear retention of defective RNAs. MATR3 forms part of complex consisting of SFPQ, NONO and MATR3.The protein contains 1 matrin type zinc finger and 2 RRM (RNA recognition motif) domains. Two transcript variants encoding the same protein have been identified for this gene.
Type: Primary
Antigen: Matrin 3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 421-520/847
Host: Rabbit
Isotype: IgG
Reactivity: Rat