Anti-HMGCS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
76079-672
: Bioss
- Antibody Type:Primary
- Antigen Name:HMGCS1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µl
- Epitope:41-150/520
- Form:Liquid
- Gene ID:266977
- Antigen Synonyms:HMCS1_HUMAN|HMGCS|3 hydroxy 3 methylglutaryl Coenzyme A synthase 1 soluble|MGC90332: EC 2.3.3.10|HMG CoA synthase|Hydroxymethylglutaryl CoA synthase, cytoplasmic
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:57 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HMGCS1
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76079-672
- Packaging:Vial
This enzyme condenses acetyl CoA with acetoacetyl CoA to form HMG CoA, which is the substrate for HMG CoA reductase.
Type: Primary
Antigen: HMGCS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 41-150/520
Host: Rabbit
Isotype: IgG
Reactivity: Mouse