Anti-eIF4E Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # 76079-430
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:eIF4E
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µl
- Epitope:121-217/217
- Form:Liquid
- Gene ID:1977
- Antigen Synonyms:Eukaryotic translation initiation factor 4E|eIF-4F 25 kDa subunit|EIF4EL1|AUTS19|EIF4E1|eIF-4E|CBP|eIF4E|EIF4F|mRNA cap-binding protein
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:24 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human eIF4E
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76079-430
- Packaging:Vial
Specifications
About this item
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures. Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit mediates the binding to the mRNA cap.
Type: Primary
Antigen: eIF4E
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 121-217/217
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat