Anti-CDKN2C Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # 76078-484
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:CDKN2C
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Epitope:101-168/168
- Form:Liquid
- Gene ID:1031
- Antigen Synonyms:Cyclin dependent kinase 6 inhibitor p18|CDN2C_HUMAN|CDKN 2C|Cyclin dependent kinase 4 inhibitor C|p18-INK4C|p18 INK6|CDK6 inhibitor p18|Cyclin dependent kinase inhibitor 2C p18 inhibits CDK4|p18 inhibits CDK4|Cyclin-dependent kinase 6 inhibitor|Cyclin dependent kinase 6 inhibitor|INK4C|Cyclin dependent kinase inhibitor 2C|Cyclin-dependent kinase 4 inhibitor C|CDKN6|CDKN2C|p18-INK6|CDKN 6|p18|OTTHUMP00000046546|p18 INK4C|Cyclin dependent inhibitor
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:18 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CDKN2C
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76078-484
- Packaging:Vial
Specifications
About this item
p18 INK4C is a specific inhibitor of cdk4 / cdk6 (it interacts strongly with CDK6 and weakly with CDK4), and is expressed during G1 to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S phase. Highest expression levels are found in skeletal muscle, but it is also found in the pancreas and heart.
Type: Primary
Antigen: CDKN2C
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-168/168
Host: Rabbit
Isotype: IgG
Reactivity: