Specifications
- Antibody Type:Primary
- Antigen Name:Surface Layer Protein
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Bacteria
- Size:100 µl
- Epitope:301-400/444
- Form:Liquid
- Gene ID:3252774
- Antigen Synonyms:SLAP_LACAC|Surface layer|S-layer protein|slpA|S-layer|surface layer protein|slpH|Slp|Lactobacillus surface layer protein
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:50 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from L, acidophilus S-layer protein
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76084-984
- Packaging:Vial
Specifications
About this item
One of the most fascinating cell envelope structures in prokaryotic organisms is two-dimensional arrays of protein or glycoprotein subunits, termed S-layers (Surface Layers). They are composed of numerous identical subunits forming a symmetric, porous, lattice-like layer that completely covers the cell surface. The subunits are held together and attached to cell wall carbohydrates by non-covalent interactions, and they spontaneously reassemble in vitro by an entropy-driven process. In lactobacilli, S-layer proteins have been detected on many but not all species. Lactobacillus S-layer proteins differ from those of other bacteria in their smaller size and high predicted pI. The positive charge in Lactobacillus S-layer proteins is concentrated in the more conserved cell wall binding domain, which can be either N- or C-terminal depending on the species. The more variable domain is responsible for the self-assembly of the monomers to a periodic structure. The biological functions of Lactobacillus S-layer proteins are poorly understood, but in some species S-layer proteins mediate bacterial adherence to host cells or extracellular matrix proteins or have protective or enzymatic functions. Lactobacillus S-layer proteins show potential for use as antigen carriers in live oral vaccine design because of their adhesive and immunomodulatory properties and the general non-pathogenicity of the species.
Type: Primary
Antigen: Surface Layer Protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/444
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria