Anti-PAX7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
76102-026
: Bioss
- Antibody Type:Primary
- Antigen Name:PAX7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Sheep,Other,Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Epitope:401-500/520
- Form:Liquid
- Gene ID:5081
- Antigen Synonyms:RMS2|HUP1|PAX7B|PAX7|Paired box protein Pax-7
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:55 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PAX7
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76102-026
- Packaging:Vial
Transcription factor playing a role in myogenesis through regulation of muscle precursor cells proliferation.
Type: Primary
Antigen: PAX7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-500/520
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Sheep; Others