Anti-Cancer testis antigen 65 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # 76118-664
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Cancer testis antigen 65
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:100 µl
- Epitope:301-400/520
- Form:Liquid
- Gene ID:7288
- Antigen Synonyms:TULP2_HUMAN|Protein P4-6|Cancer testis antigen 65|Tubby related protein 2|TUBL2|Cancer/testis antigen 65|CT65|MGC107290|Pdet|Tubby like protein 2|TUBL 2|TULP 2
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:59 kDa
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TULP2/Cancer testis antigen 65
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76118-664
- Packaging:Vial
Specifications
About this item
TULP2 is a member of a family of tubby-like (TULP) proteins which share a conserved C terminal region of approximately 200 amino acid residues. It is strongly expressed in testis and is expressed in the retina. It is also expressed in cancer cell lines.
Type: Primary
Antigen: Cancer testis antigen 65
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/520
Host: Rabbit
Isotype: IgG
Reactivity: