Anti-PTGER1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # 76118-088
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:PTGER1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Epitope:61-160/402
- Form:Liquid
- Gene ID:5731
- Antigen Synonyms:EP1|PGE receptor EP1 subtype|PTGER1|PGE2 receptor EP1 subtype|Prostanoid EP1 receptor|Prostaglandin E2 receptor EP1 subtype
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:42 kDa
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Prostaglandin E Receptor EP1
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76118-088
- Packaging:Vial
Specifications
About this item
Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(q) proteins which activate a phosphatidylinositol-calcium second messenger system. May play a role as an important modulator of renal function. Implicated the smooth muscle contractile response to PGE2 in various tissues.
Type: Primary
Antigen: PTGER1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 61-160/402
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat