- Antibody Type:Primary
- Antigen Name:TMPRSS2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Epitope:301-400/492
- Form:Liquid
- Gene ID:7113
- Antigen Synonyms:TMPRSS2|PRSS10|Transmembrane protease serine 2|PP9284|Serine protease 10
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:54 kDa
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TMPRSS2
- Purification:Purified by Protein A
- Cat. No.:76118-058
- Packaging:Vial
Serine protease that proteolytically cleaves and activates the viral spike glycoproteins which facilitate virus-cell membrane fusions; spike proteins are synthesized and maintained in precursor intermediate folding states and proteolysis permits the refolding and energy release required to create stable virus-cell linkages and membrane coalescence. Facilitates human SARS coronavirus (SARS-CoV) infection via two independent mechanisms, proteolytic cleavage of ACE2, which might promote viral uptake, and cleavage of coronavirus spike glycoprotein which activates the glycoprotein for cathepsin L-independent host cell entry. Proteolytically cleaves and activates the spike glycoproteins of human coronavirus 229E (HCoV-229E) and human coronavirus EMC (HCoV-EMC) and the fusion glycoproteins F0 of Sendai virus (SeV), human metapneumovirus (HMPV), human parainfluenza 1, 2, 3, 4a and 4b viruses (HPIV). Essential for spread and pathogenesis of influenza A virus (strains H1N1, H3N2 and H7N9); involved in proteolytic cleavage and activation of hemagglutinin (HA) protein which is essential for viral infectivity.
Type: Primary
Antigen: TMPRSS2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-400/492
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat