Anti-WNT4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # 76117-110
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:WNT4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Epitope:201-300/351
- Form:Liquid
- Gene ID:54361
- Antigen Synonyms:WNT4|UNQ426/PRO864|Protein Wnt-4|WNT-4|SERKAL
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:37 kDa
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human WNT4
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76117-110
- Packaging:Vial
Specifications
About this item
Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters (By similarity). Overexpression may be associated with abnormal proliferation in human breast tissue.
Type: Primary
Antigen: WNT4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/351
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat