Anti-SASH1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
76117-740
: Bioss
- Antibody Type:Primary
- Antigen Name:SASH1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 µl
- Epitope:421-520/1247
- Form:Liquid
- Gene ID:23328
- Antigen Synonyms:SAM and SH3 domain containing 1|SASH1_HUMAN|PEPE 1|PEPE1|SAM and SH3 domain-containing protein 1|SAM and SH3 domain containing protein 1|Proline-glutamate repeat-containing protein|RP3 323M4.1|Proline glutamate repeat containing protein|SASH 1|Sash1
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:137 kDa
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SASH1
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76117-740
- Packaging:Vial
SASH1.
Type: Primary
Antigen: SASH1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 421-520/1247
Host: Rabbit
Isotype: IgG
Reactivity: Rat