Anti-GPCR RTA Rabbit Polyclonal Antibody (FITC)
Catalog # 10315-308
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:GPCR RTA
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Epitope:51-150/343
- Form:Liquid
- Antigen Synonyms:GPR168|mrgF|RTA|MRGPRF|MGC21621|GPR140|MRGRF_HUMAN
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:38 kDa
- Storage Temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GPCR RTA
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:10315-308
- Packaging:Vial
Specifications
About this item
GPCR RTA.
Type: Primary
Antigen: GPCR RTA
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 51-150/343
Host: Rabbit
Isotype: IgG
Reactivity: