ECG Electrodes
About this item
The Radnoti 140155 ECG electrode set for mono phasic action potentials during isolated perfused heart experiments.
- Designed for ease of use
- Spring loaded platinum tipped lead
- The second lead tip is of malleable platinum wire
Spring loaded platinum tipped lead to maintain constant contact with the beating heart without destroying or damaging tissue or cells as with suction type or impaled lead type electrode sets. The second lead tip is of malleable platinum wire for ease of wrapping around aorta or aortic cannula (steel version only).
For placement under the Radnoti high tech heart chamber lid via ball and swivel clips or in open top chambers.
Each electrode lead has coaxial wire with ground for connection to bioamplifer. Shielded coaxial wire reduces noise artifacts during recordings.
2 Options Available Below
Product Details & Documents
The Radnoti 140155 ECG electrode set for mono phasic action potentials during isolated perfused heart experiments.
- Designed for ease of use
- Spring loaded platinum tipped lead
- The second lead tip is of malleable platinum wire
Spring loaded platinum tipped lead to maintain constant contact with the beating heart without destroying or damaging tissue or cells as with suction type or impaled lead type electrode sets. The second lead tip is of malleable platinum wire for ease of wrapping around aorta or aortic cannula (steel version only).
For placement under the Radnoti high tech heart chamber lid via ball and swivel clips or in open top chambers.
Each electrode lead has coaxial wire with ground for connection to bioamplifer. Shielded coaxial wire reduces noise artifacts during recordings.
Product Family Options
Product Information
- AvailabilityPrice
- MSPP-140155-MECG electrodes for mouse heart chamberSupplier Number: 140155-MSpecifications:MarketsourceMarketsource
- MSPP-140155ECG electrodes for rat heart chamberSupplier Number: 140155Specifications:MarketsourceMarketsource