Orders LinkContactUsLinkComponent
Lighthouse Canister Wall Bracket
Canister wall bracket, White
  76716-816
 :  TMWB446000
Lighthouse Canister Wall Bracket
  76716-816
Lighthouse Canister Wall Bracket
Lighthouse Canister Wall Bracket
Canister wall bracket, White
$15.82
Each
Holds surface wipe canisters securely and conveniently on mobile work stations.
  • Heavy-duty ³/₁₆" thick steel with white powdercoat finish for maximum durability
  • Integral antimicrobial coating
  • Mounts on the lower, wider pole of cart
  • Can be mounted to the upper pole with the undermounting kit
Bracket is designed to accommodate a surface wipe canister, for use on mobile work stations. The heavy-duty ³/₁₆" thick steel with white powdercoat finish provides maximum durability, and features an integral antimicrobial coating.
  • Color:
    White
  • Description:
    Canister wall bracket