Orders LinkContactUsLinkComponent
Lighthouse Scout Electrostatic Backpack Sprayer
Lighthouse Scout Electrostatic Backpack Sprayer
Scout electrostatic backpack sprayer, 2 gal.
Catalog # 76716-830
Supplier:   Unimed-Midwest
Supplier #:  SCOUTES200
Lighthouse Scout Electrostatic Backpack Sprayer
Catalog # 76716-830
Supplier:  Unimed-Midwest
Lighthouse Scout Electrostatic Backpack Sprayer
Lighthouse Scout Electrostatic Backpack Sprayer
Scout electrostatic backpack sprayer, 2 gal.
Price
$5,080.45
Log in for availability
Each
Product Details & Documents
High-efficiency touchless sprayer / disinfectant applicator and operational intelligence platform.
  • In-field service design delivers maximum uptime and reliability
  • Integrated data capture, alerts, and reporting platform drive operational intelligence and improvements
  • High-efficiency sprayer saves time and reduces chemical usage
  • Tool-free nozzle changes
  • Pump replacement in under 10 minutes with kit
The Scout ES200 delivers best-in-class efficiency, reliability, and performance to maximize cleaning and disinfecting efforts in any environment. Powered by Lighthouse Connect, Scout lets users capture, report, and leverage key disinfection data to optimize the efficiency and efficacy of infection prevention programs.
Delivery information: Supplied with:
• 2 Gallon Chemistry & Rinse Tank
• 4’ Electrostatic-Fluid Hose
• Scout ES200 Spray Wand
• 18” Spray Nozzle Extender
• 2x 75 Micron Spray Nozzles
• 2x Battery: 1 in Sprayer, 1 Spare
• Battery Charger and Cord
• Operator Grounding Wrist Strap
Specifications
  • Description:
    Scout electrostatic backpack sprayer, 2 gal.