ContactUsLinkComponent
Lighthouse Chemo Spill Cleanup Kit
Lighthouse Chemo Spill Cleanup Kit
Catalog # 76716-804
Supplier:  Unimed-Midwest
undefined
Lighthouse Chemo Spill Cleanup Kit
Catalog # 76716-804
Supplier:  Unimed-Midwest
Supplier Number:  KIT-CW
Specifications
  • Description:
    Chemo spill cleanup kit
  • Cat. No.:
    76716-804
Specifications
About this item
Designed for quick and safe cleanup of Chemotherapy drug spills.
  • Includes heavy-duty chemo-grade PPE for full protection
  • Cleanup supplies to take care of liquids
  • Super Sani-Cloth surface disinfectant wipe with 2 minute contact time to eliminate pathogens
  • Heavy-duty, extra-thick chemo waste bag for additional safety
  • Helps maintain OSHA compliance and patient safety
  • Easy to use and store, in clear zip bag, with instructions included
Kit includes: (1) Chemo Bio-Wipe bag, (1) Chemo Bloc® Maximum Protection gown, (3) absorbent towelettes, (1) pair of Chemo-Nitrile gloves, (1) fluid impermeable mask with safety shield, (1) yellow Chemo waste bag with twist tie, (1) Super Sani-Cloth disinfectant wipe, (1) Sani-Hands antibacterial hand towelette, and (1) set of instructions in English and Spanish.