To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Tumor protein p53
- Antigen Symbol:P53
- Clonality:Monoclonal
- Clone:TM-5C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:7157
- Antigen Synonyms:P53|BCC7|TRP53|LFS1
- Storage Buffer:10 mM phosphate buffer, 150 mM NaCl, pH 7.2 (50% glycerol)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic acetylated peptide (conjugated with KLH) corresponding to residues surrounding Lys382 of human TP53.
- Cat. No.:10687-296
- Supplier no.:MAB0018
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of acetylated histones and TP53.
Recommended dilutions: Western blot: 1 to 2 µg/ml; Immunofluorescence: 5 to 10 µg/ml; ELISA: 0.5 to 1 µg/ml
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: TM-5C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human