ContactUsLinkComponent
Experimentation Kits, Dot Blot Analysis
Experimentation Kits, Dot Blot Analysis
Catalog # 76286-340
Supplier:  G-Biosciences
Experimentation Kits, Dot Blot Analysis
Catalog # 76286-340
Supplier:  G-Biosciences
Supplier Number:  BE-502
Specifications
  • Description:
    Dot blot analysis, protein diagnostics
  • Cat. No.:
    76286-340
Specifications
About this item
Dot blotting is a simple technique to identify a known protein in a biological sample. The ease and simplicity of the technique makes dot blotting an ideal diagnostic tool.
  • Simulated test samples
  • Teaches immunodetection technique
  • Hands-on dot blot analysis technique with necessary reagents and supplies
The method involves applying small volumes of antigens or test samples on protein binding membrane; the membrane captures and immobilizes proteins. The proteins immobilized on the membrane are probed with a specific antibody and a matching specific detection reagent. In this lab activity students screen test samples from a population and identify those who are carriers of a disease. Students learn how dot blotting applied in the real world, for example in clinical testing, forensic, and research laboratories.
Delivery: Supplied with components needed for hands-on experimentation for six workstations of 4-5 students or 24-30 students. Supplied with teacher’s guide and separate student’s guides.