To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Eukaryotic translation initiation factor 2-alpha kinase 3
- Antigen Symbol:EIF2AK3
- Clonality:Monoclonal
- Clone:5G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:9451
- Antigen Synonyms:WRS|PEK|PERK
- Storage Buffer:0.03% sodium azide
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human EIF2AK3.
- Cat. No.:10711-766
- Supplier no.:MAB10632
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant EIF2AK3.
Recommended dilutions: ELISA: 1:10000; Western blot: 1:500 to 1:2000
Type: Primary
Antigen: EIF2AK3
Clonality: Monoclonal
Clone: 5G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human