Anti-MBD1 Rabbit Polyclonal Antibody
Catalog # 10685-290
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Methyl-CpG binding domain protein 1
- Antigen Symbol:MBD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Size:50 μg
- Cross Adsorption:No
- ChIP:Yes
- Format:Liquid
- Gene ID:4152
- Antigen Synonyms:CXXC3|PCM1|RFT
- Storage Buffer:PBS (0.05% sodium azide, 0.05% procl300)
- Storage Temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human MBD1.
- Cat. No.:10685-290
- Supplier no.:PAB14126
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MBD1.
Recommended Dilutions: ELISA: 1:300-1:1000 ChIP: 1,7 ?g/ChIP)
Type: Primary
Antigen: MBD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human