ContactUsLinkComponent
FLIPR® Membrane Potential Assay Kit
FLIPR® Membrane Potential Assay Kit
Catalog # MSPP-R8126
CAS Number:  
undefined
FLIPR® Membrane Potential Assay Kit
Catalog # MSPP-R8126
Supplier Number:  R8126
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    FLIPR membrane potential red bulk explorer kit, sufficient for 10 plates (96 or 384 wells)
  • Size:
    1 kit
  • Cat. No.:
    MSPP-R8126
Specifications
About this item
The FLIPR® Membrane Potential Assay Kit provide a fast, simple and reliable, fluorescence-based assay for detecting changes in voltage across the cell membrane and its homogenous fluorescence-based formulations for observation of real-time membrane potential changes associated with ion channel activation and ion transporter proteins.
  • More reproducible data with faster response time
  • No cumbersome pre-assay preparation
  • Ease of use at room to physiological temperatures
  • Fewer steps in the assay resulting in higher sample throughput
Each homµgeneous assay kit utilises a proprietary indicator dye and quencher combination to maximise cell line/channel/compound applicability, while eliminatiµg causes of data variability. This unique formula responds 10 times faster and hasµgreater temperature stability than traditional dyes, providiµg hµgh quality screeniµg data that showsµgood correlation with manual patch clamp assays.
Ion channel activity is highly sensitive and potentially impacted by subtle chemical changes, two FLIPR® Membrane Potential Assay Kits (Red and Blue) are available to select the optimal conditions for your delicate ion channel targets. Both formulations utilise Molecular Devices proprietary quench technology to enhance signal windows and yield acceptable Z-scores to screen a variety of targets, including TRP, ligand-, cyclic nucleotide- and voltage-gated channels.
Delivery: Explorer kit includes 10 vials of component A and one bottle of component B 1X buffer (1X HBSS + Ca²⁺, Mg²⁺, 20 mM HEPES, pH 7,4). Bulk kit includes 10 vials of component A and one bottle of component B 10X buffer, and Evaluation kit includes 5 vials of membrane potential assay Blue (Component A), 5 vials of membrane potential assay red (component A) and one bottle of component B 1X buffer.