ContactUsLinkComponent
EarlyTox™ Cardiotoxicity Kit
EarlyTox™ Cardiotoxicity Kit
Catalog # MSPP-R8210
CAS Number:  
undefined
EarlyTox™ Cardiotoxicity Kit
Catalog # MSPP-R8210
Supplier Number:  R8210
CAS Number:  
Specifications
  • Description:
    EarlyTox™ cardiotoxicity explorer kit
  • Size:
    1 kit
  • Cat. No.:
    MSPP-R8210
  • Includes:
    2 vials component A, each vial sufficient for assay of one 96 or 384 well plate (total 2 plates/kit), plus 1 bottle of component B (dilution buffer) and one vial each of 3 reference compounds: isoproterenol, sotalol, and propranolol
  • Supplier no.:
    R8210
Specifications
About this item
The EarlyTox™ Cardiotoxicity Kit provides a fast, simple and reliable fluorescence based method for identifying cardiotoxic compounds in a biorelevant assay using stem-cell derived cardiomyocytes.
  • Eliminate cardiotoxic compounds and identify potential drug candidates
  • Measure 384 samples in minutes rather than hours
  • Minimise non specific effect of dye on beat characteristics
  • Reduce well-to-well variation and improve data quality
  • Largest signal dynamic range available
  • Benchmark against three reference compounds
For researchers lookiµg to identify cardiotoxic compounds, the EarlyTox™ Cardiotoxicity Kit allows screeniµg of more compounds earlier in drµg discovery. This makes it possible to prioritise leads and direct medicinal chemistry efforts sooner, improviµg productivity and reduciµg costs associated with extensive safety testing downstream.