Human Putative pre-mRNA-splicing Factor ATP-dependent RNA Helicase DHX32(DHX32) ELISA Kit
Catalog # 76702-758
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:Putative pre-mRNA-splicing Factor ATP-dependent RNA helicase DHX32
- Description:Human Putative pre-mRNA-splicing Factor ATP-dependent RNA helicase DHX32(DHX32) ELISA Kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:62.5 - 2000 pg/ml
- Storage Temperature:2…8 °C
- Application:Signal Transduction
- Sample Volume:50 - 100 µl
- Sensitivity:7.8 pg/ml
- Regulatory Status:RUO
- Cat. No.:76702-758
Specifications
About this item
- Detection wavelength: 450 nm