Human Hepatitis B Virus X Ag(HBxAg) ELISA Kit
Catalog # 76703-188
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:Hepatitis B virus X Ag
- Description:Human hepatitis B virus X Ag(HBxAg) ELISA Kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Qualitative
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:Positive / Negative
- Storage Temperature:2…8 °C
- Application:Microbiology
- Sample Volume:50 - 100 µl
- Sensitivity:Positive/Negative
- Regulatory Status:RUO
- Cat. No.:76703-188
Specifications
About this item
- Detection wavelength: 450 nm