Human Hepatic Lipase(HL) ELISA Kit
Catalog # 76713-156
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:Hepatic lipase
- Description:Human hepatic lipase(HL) ELISA Kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:16 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Metabolism
- Sample Volume:50 - 100 µl
- Sensitivity:4.5 pg/ml
- Regulatory Status:RUO
- Cat. No.:76713-156
Specifications
About this item
- Detection wavelength: 450 nm