Human Low affinity Immunoglobulin gamma Fc region Receptor II-c(FCGR2C) ELISA Kit
Catalog # 76700-132
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:Low affinity immunoglobulin gamma Fc region receptor II-c
- Description:Human Low affinity immunoglobulin gamma Fc region receptor II-c(FCGR2C) ELISA Kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Storage Temperature:2…8 °C
- Application:Immunology
- Sample Volume:50 - 100 µl
- Regulatory Status:RUO
- Cat. No.:76700-132
Specifications
About this item
- Detection wavelength: 450 nm