Human Methionine Adenosyltransferase 2 Subunit beta (MAT2B) ELISA Kit
76703-052
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:Methionine adenosyltransferase 2 subunit beta
- Description:Human Methionine adenosyltransferase 2 subunit beta(MAT2B) ELISA Kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (−20 °C)
- Detection Range:37.5 - 1200 pg/ml
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:3 pg/ml
- Regulatory Status:RUO
- Cat. No.:76703-052
- Detection wavelength: 450 nm