ContactUsLinkComponent
Heavy-Duty Flask Tongs
Heavy-Duty Flask Tongs
Catalog # 470344-264
Supplier:  Wards
undefined
Heavy-Duty Flask Tongs
Catalog # 470344-264
Supplier:  Wards
Supplier Number:  CH0733
Specifications
  • Description:
    Heavy-duty flask tongs
  • Length:
    10"
  • Environmentally Preferable:
  • Cat. No.:
    470344-264
Specifications
About this item
Made of stainless steel and measuring 10" long, these tongs with cork-lined jaws offer a strong grip for secure handling of flasks in laboratory settings.
  • Heavy-duty flask tongs made from stainless steel
  • Square opening for miscellaneous applications
  • Cork-lined jaws to use with hot flasks and glassware
  • Permits handling of hot flasks and objects
Heavy-duty flask tongs feature a cork-lined jaw for handling hot flasks and glassware and have a square opening for miscellaneous applications. Permits handling of hot flasks and glassware. The flask tongs measure approximately 10" long and 4.25" wide when closed. Reliable heavy-duty build and design.