ContactUsLinkComponent
Heavy Duty Multi-Purpose Table Clamp
  470345-290
 :  Wards
undefined
Heavy Duty Multi-Purpose Table Clamp
  470345-290
 :  Wards
 :  CH0688FBG
  • Description:
    Heavy duty multi-purpose table clamp
  • Cat. No.:
    470345-290
Made of die-cast metal, this clamp features a vinyl-coated grip and swivel pad, along with a built-in rod and pulley holder for versatile applications.
  • High performance, heavy duty multi-purpose table clamp
  • It's versatile design allows you to secure a wide variety of laboratory apparatus and accessories
  • Non-ferrous, drop forged steel construction that will not bend or break under heavy loads
  • Convenient built-in rod or pulley holder accommodates rods and pulleys up to 16 mm
  • Features a vinyl sleeve and swivel pad to ensure a strong, slip-proof grip on a variety of surfaces
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms
Can be used for holding rods and pulleys, immobilizing meter sticks, securing lab equipment, and can also function as a G-Clamp. Made from heavy duty, die-cast metal, and features a vinyl sleeve and swivel pad to ensure a strong, slip proof grip on a variety of surfaces, up to 65 mm (2.5") thick. Non-ferrous, drop forged steel construction that will not bend or break under heavy loads. Solid metal screw adjustments and handles won't break while tightening. The built-in holder can accommodate rods or pulleys measuring up to 16 mm in diameter. Clamp measures approximately 5" wide, 4"tall and 1.5" long.