Platinum Wire
Platinum wire
Catalog # 470344-750
Supplier: Wards
Supplier #: CH0538
Jump to:
Product Details & Documents
Platinum wire supplied in a glass tube, designed specifically for flame tests, ensuring accurate and consistent results.
- Platinum wire ideal for flame test
- Supplied in borosilicate glass tube, fused into glass handle
- Attached to silicon cork
- Tube measures 5" long and 0.6" in diameter. Wire measures 18 to 20 mm long and approximately 0.12 mm in diameter
Specifications
- Description:Platinum wire