Order Entry
Puerto Rico
ContactUsLinkComponent
 
Economy Hand Crank Centrifuge with Tubes
  470346-058
 :  Wards
undefined
Economy Hand Crank Centrifuge with Tubes
  470346-058
 :  Wards
 :  CH0267

 

  • Description:
    Economy hand crank centrifuge with tubes
  • Cat. No.:
    470346-058

 

 

Hand crank centrifuge capable of holding 4×15 ml tubes, includes tubes for immediate use.

  • Single speed hand crank centrifuge consisting of die casted parts, a steel pinion, and worm drive
  • Features a removable crank handle
  • Stands 9" tall and is 4" wide
 : Supplied with four polythene tubes.