ContactUsLinkComponent
Burette/Tube Clamp
Burette/Tube Clamp
Catalog # 470344-546
Supplier:  Wards
undefined
Burette/Tube Clamp
Catalog # 470344-546
Supplier:  Wards
Supplier Number:  CH0258A
Specifications
  • Description:
    Burette/tube clamp
  • Cat. No.:
    470344-546
Specifications
About this item
Single burette/tube clamp made of high-strength alloy, equipped with spring-loaded jaws, has a width capacity of ⁷/₈", holds burettes up to 100 ml, and fits maximum rod diameter of 16 mm.
  • Spring loaded, high strength alloy jaws holds objects in place
  • Powder coated for long lasting finish
This burette clamp made from high strength alloy has spring-loaded jaws suitable for all burettes up to 100 ml. Can be mounted on rods up to 16 mm diameter.