ContactUsLinkComponent
Latex Pipette Bulb
Latex Pipette Bulb
Catalog # 470345-386
Supplier:  Wards
undefined
Latex Pipette Bulb
Catalog # 470345-386
Supplier:  Wards
Supplier Number:  CH0232C
Specifications
  • Description:
    Latex pipette bulb
  • Cat. No.:
    470345-386
Specifications
About this item
Latex pipette bulb with a diameter range of 6 to 8 mm and a maximum capacity of 1 ml.