Rat Adisintegrin and Aetalloproteinase with Thrombospondin Motifs 5(ADAMTS5) ELISA Kit
Catalog # 76713-706
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:Adisintegrin and metalloproteinase with thrombospondin motifs 5
- Description:Rat adisintegrin and metalloproteinase with thrombospondin motifs 5(ADAMTS5) ELISA kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (−20 °C)
- Detection Range:80 - 4000 pg/ml
- Storage Temperature:2…8 °C
- Application:Cancer, cardiovascular
- Sample Volume:50 - 100 μl
- Sensitivity:25 pg/ml
- Regulatory Status:RUO
- Cat. No.:76713-706
Specifications
About this item
- Detection wavelength: 450 nm