Rat Adenosine Monophosphate (AMP) ELISA Kit
Catalog # 76710-252
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:Adenosine monophosphate
- Description:Rat adenosine monophosphate (AMP) ELISA kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (−20 °C)
- Detection Range:16 - 1000 nmol/L
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 μl
- Sensitivity:6.5 nmol/L
- Regulatory Status:RUO
- Cat. No.:76710-252
Specifications
About this item
- Detection wavelength: 450 nm