Human Chondroitin Sulfate-846 (CS-846) ELISA Kit
Catalog # 76715-014
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:Chondroitin sulfate-846
- Description:Human chondroitin sulfate-846 (CS-846) ELISA kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (−20 °C)
- Storage Temperature:2…8 °C
- Application:Cardiovascular
- Sample Volume:50 - 100 μl
- Regulatory Status:RUO
- Cat. No.:76715-014
Specifications
About this item
- Detection wavelength: 450 nm