ContactUsLinkComponent
Porcelain Combustion Boat with Handle
Porcelain Combustion Boat with Handle
Catalog # 470346-002
Supplier:  Wards
undefined
Porcelain Combustion Boat with Handle
Catalog # 470346-002
Supplier:  Wards
Supplier Number:  CH0751
Specifications
  • Description:
    Porcelain combustion boat with handle
  • Cat. No.:
    470346-002
Specifications
About this item
Measuring 4×1" exterior and 3×¹/₂" interior, these porcelain boats with handles come in a pack of 10 and provide a reliable container for combustion experiments.
  • Pack of 10 porcelain combustion boats with handle
  • Made from porcelain for increased resistance to heat and caustic chemicals
  • Interior measures approximately 3×1"
  • Each pack comes with 10 individual porcelain combustion boats
  • Great for any classroom or laboratory