Hoffman Electrolysis Metal Stand
Catalog # 470344-862
Supplier: Wards
Specifications
- Description:Hoffman electrolysis metal stand
- Cat. No.:470344-862
Specifications
About this item
Measuring 24" in overall height, this metal stand with support clamps provides stability and versatility for Hoffman electrolysis experiments.
- 8×5" base is powder-coated
- 23.6" stainless steel rod
Hoffman Electrolysis Metal stand with support clamps is designed to hold components for Hoffman electrolysis equipment.