Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Sampling Tongs

Sampling Tongs

Sampling tongs, 8.25"
Catalog # 470345-158
Supplier:   Wards
Supplier #:  CH0736
Sampling Tongs
Catalog # 470345-158
Supplier:  Wards
Sampling Tongs
Sampling Tongs

Sampling tongs, 8.25"

Price
$6.79
Log in for availability
Each
Product Details & Documents

These nickel-plated tongs are ideal for sampling purposes, providing a reliable and precise grip for various materials in laboratory and industrial environments.

  • 8.25" long sampling spoon/tongs
  • 1" spoon splits in half vertically; useful for collecting hot samples of thick materials
  • Designed to handle mercury safely
  • Scalloped spoon design; half scallop splits into a quarter per side
Specifications
  • Description:
    Sampling tongs
  • Length:
    8.25"