Order Entry
Puerto Rico
ContactUsLinkComponent
Crucible Tongs with Straight Tips
Crucible Tongs with Straight Tips
Catalog # 470344-964
Supplier:  Wards
undefined
Crucible Tongs with Straight Tips
Catalog # 470344-964
Supplier:  Wards
Supplier Number:  CH0732B

Specifications

  • Description:
    Crucible tongs with straight tips
  • Length:
    12"
  • Cat. No.:
    470344-964

Specifications

About this item

Measuring 12" long, these stainless steel tongs features a bow design, straight serrated tips, and are suitable for various laboratory applications.

  • Crucible tongs made of high-quality, durable stainless steel
  • Straight serrated tips
  • Tongs measure 12" long and open 9" wide

Crucible tongs made of high-quality, durable stainless steel are useful for grasping hot crucibles, flasks, beakers, or dishes. Perfect for any classroom or laboratory.