Order Entry
United States
ContactUsLinkComponent
 
Crucible Tongs with Straight Tips
  470343-950
 :  Wards
undefined
Crucible Tongs with Straight Tips
  470343-950
 :  Wards
 :  CH0729B

 

  • Description:
    Crucible tongs with straight tips
  • Length:
    9.5"
  • Environmentally Preferable:
  • Cat. No.:
    470343-950

 

 

These metal tongs feature a bow design and straight serrated tips. They provide a secure grip for crucibles in laboratory settings.

  • General-purpose, metal crucible tongs
  • Straight, serrated tips and bow
  • Tongs measure 9.5" in length
  • Perfect for any classroom or laboratory

General-purpose metal crucible tongs with small bow. Straight, serrated tips. Useful for grasping hot crucibles, evaporating dishes, or small flasks and beakers. Tongs measure 9.5" in total length, and 3.25" wide when closed. Each tip is approximately 0.9" long, and bow can hold items with a minimum diameter of 1".