ContactUsLinkComponent
2-Prong Universal Retort Clamp
2-Prong Universal Retort Clamp
Catalog # 470344-468
Supplier:  Wards
undefined
2-Prong Universal Retort Clamp
Catalog # 470344-468
Supplier:  Wards
Supplier Number:  CH0688D
Specifications
  • Description:
    2-prong universal retort clamp
  • Cat. No.:
    470344-468
Specifications
About this item
The 2-prong, double adjustable universal clamp offers a non-slip vinyl-coated grip, swivel bosshead, and is made of durable stainless steel.
  • High performance, die casted metal retort clamp, suitable for holding a variety of items. The clamp features two, vinyl coated prongs that are specifically designed for maximum traction and grip. Ideal for use with items that have straight sides
  • Clamp features adjustable thumbscrews on both sides, allowing for a wider range of accommodation
  • Clamp is attached to swivel bosshead that fits rods measuring up to 21 mm in diameter
  • Designed for use in professional laboratories, but is an ideal choice for science classroom
This jumbo size, universal clamp is designed to hold a variety of items. The two, vinyl coated jaws provide a firm grip and are adjustable on both sides with thumbscrews for a wide range of accommodation. Clamp is ideal for use with items that have straight sides, such as burettes, test tubes and flask necks. Accommodates apparatus up to 2.15" (55 mm) wide. Overall length is 8".