ContactUsLinkComponent
3-Finger Lab Clamp with Bosshead
3-Finger Lab Clamp with Bosshead
Catalog # 470345-740
Supplier:  Wards
undefined
3-Finger Lab Clamp with Bosshead
Catalog # 470345-740
Supplier:  Wards
Supplier Number:  CH0688
Specifications
  • Description:
    3-finger lab clamp with bosshead
  • Cat. No.:
    470345-740
Specifications
About this item
Lab clamp with a 3-finger design, including a boss head clamp holder for lab stand rods, PVC-coated prongs capable of holding objects up to 3.5" in diameter, and additional features such as a beaker holder and flask clamp, providing versatility and convenience for various lab applications.
  • Die cast alloy construction is strong and corrosion-resistant
  • 3-finger jaws on clamp open to 3.5" (90 mm) wide for holding a wide range of glassware sizes and volumes
  • PVC-coated prongs provide a strong, slip-free hold that won’t scratch objects. PVC coating can withstand heat up to 100 °C for short periods of time but it is recommended you remove the PVC coating for extreme temperature uses
  • Swivel bosshead for rotating clamped objects 360°. Bosshead attaches to rods up to 15 mm in diameter. Clamp extends out approximately 5" from rod (not adjustable)
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms
Die casted alloy clamp retort, PVC coated jaws with swivel boss head which accommodates rods up to 15 mm in diameter. Jaws have a capacity of 3.5" or 90 mm.