ContactUsLinkComponent
Universal Clamp with Bosshead
Universal Clamp with Bosshead
  470343-672
 :  Wards
undefined
Universal Clamp with Bosshead
  470343-672
 :  Wards
 :  CH0679AB
  • Description:
    Universal clamp with bosshead
  • Cat. No.:
    470343-672
Universal clamp with cork-lined prongs capable of holding objects up to 2.3" wide, featuring a boss head for attaching to rods up to 0.59" in diameter, offering secure and adjustable holding for various lab setups.
  • Boss head holds rods up to 15 mm in diameter
  • Adjustable jaws hold objects up to 60 mm in width
  • 1" cork lined jaws
  • Perfect for test tubes, burettes, or flasks
Two prong, die casted with other parts of plated brass, accepts articles from 15 to 60 mm diameter actuated by nickel plated brass pummels and twin screw. The jaws are cork lined. Fitted with boss head.