Heavy-Duty Cork Boring Machine
Heavy-duty cork boring machine
Catalog # 470345-462
Supplier: Wards
Supplier #: CH0326A
Jump to:
Product Details & Documents
Sturdy and robust cork boring machine designed for heavy-duty usage, ensuring efficient and precise cork-boring tasks.
- 12 nickel-plated borers included
- Cutting sizes 7 to 25 mm
- Hand crank
- 4 pre-drilled holes in base for mounting to a table top
The cork boring machine is designed for drilling uniform and straight holes in cork or rubber stoppers. Cast iron frame with wood platform (10 cm diameter) for support. Adjustable height. Completed with set of 12 nickel-plated borers. Base plate has 4 mounting holes to keep boring machine stable during use.
Specifications
- Description:Heavy-duty cork boring machine