Orders LinkContactUsLinkComponent
Plastic Single Burette Clamp
Plastic single burette clamp
  470345-314
 :   Wards
 :  CH0262A
Plastic Single Burette Clamp
  470345-314
 :  Wards
Plastic Single Burette Clamp
Plastic Single Burette Clamp
Plastic single burette clamp
$4.20
Each
Single polypropylene burette clamp.
  • Single clamp - fits rods 10 to 15 mm in diameter
  • Features two tapered wedges and a spring-loaded jack
  • Made of high quality polypropylene
The sturdy, polypropylene single burette clamp is a great tool for tightly holding 10 and 12.5 mm rods. The spring-loaded jack and two tapered wedges assure a snug and safe fit.
  • Description:
    Plastic single burette clamp