Order Entry
Puerto Rico
ContactUsLinkComponent
4-Chloro-7-methyl-5,6,7,8-tetrahydropyrido[4',3':4,5]thieno[2,3-d]pyrimidine 97%
Toxic
4-Chloro-7-methyl-5,6,7,8-tetrahydropyrido[4',3':4,5]thieno[2,3-d]pyrimidine 97%
Catalog # 76678-466
Supplier:  Ambeed
CAS Number:  
undefined
Toxic
4-Chloro-7-methyl-5,6,7,8-tetrahydropyrido[4',3':4,5]thieno[2,3-d]pyrimidine 97%
Catalog # 76678-466
Supplier:  Ambeed
Supplier Number:  A1219460-1G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g
  • Cat. No.:
    76678-466

Specifications

About this item

 :
 :239.7245 g/mol
UN:2811
ADR:6.1,Ⅲ
 :GHS06
CAS Number:26830-29-5
Formula:C₁₀H₁₀ClN₃S
MDL Number:MFCD08695126